siRNA Details
Gene Symbol: RGD1305178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025003.1 | NP_001020174.1 | 5, 6 | 400 | ||
XM_006233915.2 | XP_006233977.1 | 5, 6 | 438 | |||
XM_008761665.1 | XP_008759887.1 | 5, 6 | 354 | |||
GenBank mRNA | BC091189.1 | - | 5, 6 | 400 | 7367873 |
