siRNA Details
Gene Symbol: Fam84a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127299.1 | NP_001120771.1 | 4 | 708 | ||
XM_006239901.2 | XP_006239963.1 | 2 | 360 | |||
GenBank mRNA | BC166716.1 | - | 2 | 360 | 7930888 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029007, BC005488, AK014625.
