siRNA Details
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007802.3 | NP_001007803.1 | 5 | 481 | ||
XM_006243969.2 | XP_006244031.1 | 4 | 503 | |||
GenBank mRNA | AY325262.1 | - | 5 | 481 | ||
AY387088.1 | - | 4 | 441 | |||
BC107930.1 | - | 4 | 474 | 7130158 |
