siRNA Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173840.1 | NP_776212.1 | 8, 9 | 404 | ||
XM_006249160.2 | XP_006249222.1 | 9, 10 | 575 | |||
XM_008769138.1 | XP_008767360.1 | 8, 9 | 857 | |||
XM_008769139.1 | XP_008767361.1 | 7, 8 | 821 | |||
GenBank mRNA | AY170849.1 | - | 8, 9 | 404 | ||
FQ231408.1 | - | 6, 7 | 264 | |||
FQ235223.1 | - | 6, 7 | 264 |
