siRNA Details
Gene Symbol: Ap1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108331.2 | NP_001101801.2 | 3, 4 | 376 | ||
XM_006249179.2 | XP_006249241.1 | 3, 4 | 405 | |||
GenBank mRNA | BC169070.1 | - | 3, 4 | 320 | 5598714 | |
FQ230081.1 | - | 2, 3 | 240 |
