siRNA Details
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108390.2 | NP_001101860.2 | 2, 3 | 588 | ||
XM_006252429.1 | XP_006252491.1 | 3, 4 | 533 | |||
GenBank mRNA | CO398212.1 | - | 2, 3 | 371 | ||
DY565248.1 | - | 1, 2 | 150 |
