siRNA Details
Gene Symbol: Rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014173.1 | NP_001014195.1 | 4 | 745 | ||
XM_006232686.2 | XP_006232748.1 | 3 | 577 | |||
XM_006232688.2 | XP_006232750.1 | 3 | 302 | |||
XM_006232689.2 | XP_006232751.1 | 3 | 577 | |||
XM_008761179.1 | XP_008759401.1 | 3 | 537 | |||
GenBank mRNA | BC079059.1 | - | 4 | 745 | 7115706 |
