siRNA Details
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108651.1 | NP_001102121.1 | 1, 2 | 192 | ||
XM_006237529.1 | XP_006237591.1 | 1, 2 | 201 | |||
GenBank mRNA | BC168221.1 | - | 1, 2 | 150 | 7323058 | |
FQ220508.1 | - | 1, 2 | 192 | |||
FQ229464.1 | - | 1, 2 | 192 | |||
FQ233110.1 | - | 1, 2 | 161 | |||
FQ235087.1 | - | 1, 2 | 161 |
