siRNA Details
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013224.1 | NP_001013242.1 | 5 | 729 | ||
XM_006243608.2 | XP_006243670.1 | 5 | 1073 | |||
XM_006243609.2 | XP_006243671.1 | 5 | 1077 | |||
XM_006243610.2 | XP_006243672.1 | 5 | 693 | |||
XM_008766541.1 | XP_008764763.1 | 3 | 815 | |||
GenBank mRNA | BC083725.1 | - | 5 | 729 | 7191168 |
