siRNA Details
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025745.1 | NP_001020916.1 | 6 | 759 | ||
XM_006245245.2 | XP_006245307.1 | 7 | 665 | |||
XM_006245246.2 | XP_006245308.1 | 7 | 647 | |||
GenBank mRNA | BC099215.1 | - | 6 | 759 | 7384656 |
