siRNA Details
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108815.1 | NP_001102285.1 | 3 | 407 | ||
GenBank mRNA | BC079470.1 | - | 3 | 385 | ||
BC158792.1 | - | 3 | 426 | 7312115 | ||
EF688600.1 | - | 3 | 385 |
