siRNA Details
Gene Symbol: Bloc1s4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100766.1 | NP_001094236.1 | 1 | 420 | ||
GenBank mRNA | BC091265.1 | - | 1 | 394 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bloc1s4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100766.1 | NP_001094236.1 | 1 | 420 | ||
GenBank mRNA | BC091265.1 | - | 1 | 394 |