siRNA Details
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751585.3 | XP_003751633.1 | 2 | 884 | ||
GenBank mRNA | BC088409.1 | - | 2 | 838 | 7189104 |

More Information
Other Species Targeted

