siRNA Details
Gene Symbol: Olr1445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000958.1 | NP_001000958.1 | 1 | 588 |
Gene Symbol: LOC100911586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750835.1 | XP_003750883.1 | 588 |
