siRNA Details
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031629.1 | NP_113817.1 | 4 | 553 | ||
GenBank mRNA | FQ215475.1 | - | 4 | 571 | ||
FQ218364.1 | - | 4 | 572 | |||
FQ219781.1 | - | 4 | 571 | |||
FQ220578.1 | - | 4 | 570 | |||
FQ220909.1 | - | 4 | 572 | |||
FQ225189.1 | - | 4 | 610 | |||
FQ226073.1 | - | 4 | 571 | |||
FQ226711.1 | - | 4 | 571 | |||
FQ228014.1 | - | 4 | 571 | |||
FQ232850.1 | - | 4 | 572 | |||
L17127.1 | - | 4 | 553 |
