siRNA Details
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053418.2 | NP_445870.1 | 10 | 846 | ||
XM_008768904.1 | XP_008767126.1 | 10 | 863 | |||
GenBank mRNA | AF234601.1 | - | 10 | 821 | ||
BC161818.1 | - | 10 | 857 | 9101868 |
