siRNA Details
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053418.2 | NP_445870.1 | 6, 7 | 595 | ||
XM_008768904.1 | XP_008767126.1 | 6, 7 | 612 | |||
GenBank mRNA | AF234601.1 | - | 6, 7 | 570 | ||
BC161818.1 | - | 6, 7 | 606 | 9101868 | ||
CK599711.1 | - | 595 |
