siRNA Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081958.1 | NP_001075427.1 | 15 | 1679 | ||
GenBank mRNA | CA505625.1 | - | 620 | |||
FQ223839.1 | - | 1317 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081958.1 | NP_001075427.1 | 15 | 1679 | ||
GenBank mRNA | CA505625.1 | - | 620 | |||
FQ223839.1 | - | 1317 |