siRNA Details
Gene Symbol: Cxcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102587.1 | NP_001096057.1 | 1 | 84 | ||
GenBank mRNA | DQ132631.1 | - | 1 | 84 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102587.1 | NP_001096057.1 | 1 | 84 | ||
GenBank mRNA | DQ132631.1 | - | 1 | 84 |