siRNA Details
Gene Symbol: Ceacam6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB434131.1 | - | 3 | 492 | ||
BC078962.1 | - | 3 | 731 | 7111028 |
Gene Symbol: LOC103689938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759078.1 | XP_008757300.1 | 774 |
