siRNA Details
Gene Symbol: LOC100125384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103363.1 | NP_001096833.1 | 1 | 1041 | ||
GenBank mRNA | BC098006.1 | - | 1 | 1041 | 7453927 | |
BC105848.1 | - | 1 | 1022 |
Gene Symbol: LOC103690168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759465.1 | XP_008757687.1 | 1123 |
