siRNA Details
Gene Symbol: NEWGENE_1305281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233223.2 | XP_006233285.1 | 9 | 1241 | ||
XM_006233224.2 | XP_006233286.1 | 9 | 1356 | |||
XM_008761473.1 | XP_008759695.1 | 9 | 1112 |
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106467.3 | NP_001099937.1 | 1109 | |||
GenBank mRNA | BC160912.1 | - | 1098 | 7456962 |
