siRNA Details
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126120.1 | NP_001119592.1 | 4 | 308 | ||
GenBank mRNA | BC083724.1 | - | 4 | 308 | ||
FQ212950.1 | - | 4 | 364 | |||
FQ213560.1 | - | 4 | 353 |
