siRNA Details
Gene Symbol: Gng12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053661.1 | NP_446113.1 | 4 | 247 | ||
XM_006236610.2 | XP_006236672.1 | 5 | 811 | |||
XM_006236612.2 | XP_006236674.1 | 3 | 212 | |||
XM_008762969.1 | XP_008761191.1 | 5 | 711 | |||
XM_008762970.1 | XP_008761192.1 | 5 | 686 | |||
XM_008762971.1 | XP_008761193.1 | 3 | 864 | |||
XM_008762972.1 | XP_008761194.1 | 3 | 712 | |||
GenBank mRNA | AF022091.1 | - | 2 | 86 | ||
DQ120501.1 | - | 2 | 45 | |||
DQ120502.1 | - | 2 | 45 | |||
FM031435.1 | - | 5 | 265 | |||
FQ223368.1 | - | 5 | 285 |
