siRNA Details
Gene Symbol: Gng12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053661.1 | NP_446113.1 | 3 | 174 | ||
XM_006236610.2 | XP_006236672.1 | 4 | 738 | |||
XM_006236612.2 | XP_006236674.1 | 2 | 139 | |||
XM_008762969.1 | XP_008761191.1 | 4 | 638 | |||
XM_008762970.1 | XP_008761192.1 | 4 | 613 | |||
XM_008762971.1 | XP_008761193.1 | 2 | 791 | |||
XM_008762972.1 | XP_008761194.1 | 2 | 639 | |||
GenBank mRNA | AF022091.1 | - | 1 | 13 | ||
FM031435.1 | - | 4 | 192 | |||
FQ223368.1 | - | 4 | 212 |
