siRNA Details
Gene Symbol: Akap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053665.1 | NP_446117.1 | 1 | 1304 | ||
XM_006247067.2 | XP_006247129.1 | 2 | 1477 | |||
XM_006247068.1 | XP_006247130.1 | 2 | 1473 | |||
XM_008767930.1 | XP_008766152.1 | 2 | 1532 | |||
GenBank mRNA | AF068202.1 | - | 1 | 1304 | ||
BC081753.1 | - | 2 | 1453 | |||
BC097298.1 | - | 2 | 1465 | |||
BC129090.1 | - | 2 | 1444 |
