siRNA Details
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053671.2 | NP_446123.2 | 17 | 3239 | ||
GenBank mRNA | BM390672.1 | - | 1 | 216 | ||
BQ780433.1 | - | 1 | 212 |
Gene Symbol: LOC102548101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762223.1 | XP_008760445.1 | 1 | 2019 |
