siRNA Details
Gene Symbol: Zp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053762.1 | NP_446214.1 | 5 | 826 | ||
GenBank mRNA | BC127488.1 | - | 5 | 826 | 7308805 | |
D78482.1 | - | 5 | 826 | |||
Y10823.1 | - | 5 | 819 |
