siRNA Details
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105744.2 | NP_001099214.2 | 17 | 2763 | ||
GenBank mRNA | BQ192270.1 | - | 3 | 255 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105744.2 | NP_001099214.2 | 17 | 2763 | ||
GenBank mRNA | BQ192270.1 | - | 3 | 255 |