siRNA Details
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105744.2 | NP_001099214.2 | 19 | 2873 | ||
GenBank mRNA | BQ192270.1 | - | 1 | 145 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177806, NM_013719, AF193343, AK129334, AF193342, and others.
Gene Ontology Categories:
