siRNA Details
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053817.2 | NP_446269.2 | 20, 21 | 4796 | ||
XM_008764731.1 | XP_008762953.1 | 18, 19 | 4049 | |||
XM_008764732.1 | XP_008762954.1 | 5, 6 | 3192 | |||
XM_008764733.1 | XP_008762955.1 | 5, 6 | 3195 | |||
GenBank mRNA | L14851.1 | - | 20, 21 | 4796 |
