siRNA Details
Gene Symbol: Nrxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053846.1 | NP_446298.1 | 18 | 3853 | ||
XM_008760057.1 | XP_008758279.1 | 19 | 4086 | |||
XM_008760058.1 | XP_008758280.1 | 19 | 4077 | |||
XM_008760059.1 | XP_008758281.1 | 18 | 4062 | |||
XM_008760060.1 | XP_008758282.1 | 18 | 4053 | |||
XM_008760061.1 | XP_008758283.1 | 18 | 4050 | |||
XM_008760062.1 | XP_008758284.1 | 19 | 4077 | |||
XM_008760063.1 | XP_008758285.1 | 18 | 4053 | |||
XM_008760064.1 | XP_008758286.1 | 3 | 948 | |||
GenBank mRNA | M96376.1 | - | 18 | 3853 | ||
M96377.1 | - | 3 | 928 |
