siRNA Details
Gene Symbol: Nrxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053846.1 | NP_446298.1 | 22 | 5172 | ||
XM_008760057.1 | XP_008758279.1 | 23 | 5405 | |||
XM_008760058.1 | XP_008758280.1 | 23 | 5396 | |||
XM_008760059.1 | XP_008758281.1 | 22 | 5381 | |||
XM_008760060.1 | XP_008758282.1 | 22 | 5372 | |||
XM_008760061.1 | XP_008758283.1 | 21 | 5279 | |||
XM_008760062.1 | XP_008758284.1 | 23 | 4814 | |||
XM_008760063.1 | XP_008758285.1 | 22 | 4790 | |||
XM_008760064.1 | XP_008758286.1 | 7 | 2267 | |||
GenBank mRNA | M96376.1 | - | 22 | 5172 | ||
M96377.1 | - | 7 | 2247 |
