siRNA Details
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053850.1 | NP_446302.1 | 2 | 208 | ||
XM_006234894.2 | XP_006234956.1 | 3 | 348 | |||
XM_006234895.2 | XP_006234957.1 | 3 | 333 | |||
GenBank mRNA | BC078766.1 | - | 2 | 220 | 7126157 | |
FQ213399.1 | - | 3 | 287 | |||
FQ230935.1 | - | 2 | 167 | |||
M81681.1 | - | 2 | 208 |
