siRNA Details
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751160.3 | XP_003751208.1 | 17 | 2316 | ||
XM_003751161.3 | XP_003751209.1 | 17 | 2009 | |||
XM_006249202.2 | XP_006249264.1 | 17 | 3016 | |||
XM_006249203.2 | XP_006249265.1 | 16 | 2956 | |||
XM_006249204.2 | XP_006249266.1 | 17 | 2715 | |||
XM_006249205.2 | XP_006249267.1 | 17 | 2709 | |||
XM_008769165.1 | XP_008767387.1 | 16 | 2243 | |||
XM_347163.7 | XP_347164.4 | 17 | 3022 | |||
GenBank mRNA | U09229.1 | - | 1 | 25 |
