siRNA Details
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053865.1 | NP_446317.1 | 5 | 2442 | ||
XM_008764690.1 | XP_008762912.1 | 3 | 472 | |||
GenBank mRNA | FQ212949.1 | - | 3 | 383 | ||
FQ214343.1 | - | 3 | 308 | |||
U17603.1 | - | 3 | 442 | |||
U17604.1 | - | 5 | 2442 |
