siRNA Details
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109911.1 | NP_001103381.1 | 16 | 2185 | ||
NM_053876.2 | NP_446328.2 | 17 | 2068 | |||
XM_008765198.1 | XP_008763420.1 | 14 | 1915 | |||
GenBank mRNA | AY724543.1 | - | 16 | 2172 | ||
BC088317.1 | - | 12 | 1696 | 7325202 | ||
U32577.2 | - | 17 | 2068 |
