siRNA Details
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109911.1 | NP_001103381.1 | 8, 9 | 856 | ||
NM_053876.2 | NP_446328.2 | 9, 10 | 739 | |||
XM_006241116.2 | XP_006241178.1 | 8, 9 | 2046 | |||
XM_008765198.1 | XP_008763420.1 | 6, 7 | 586 | |||
GenBank mRNA | AY724543.1 | - | 8, 9 | 843 | ||
BC088317.1 | - | 4, 5 | 367 | 7325202 | ||
EV774465.1 | - | 9, 10 | 776 | |||
U32577.2 | - | 9, 10 | 739 |
