siRNA Details
Gene Symbol: Rere | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053885.2 | NP_446337.2 | 2, 3 | 900 | ||
XM_006239438.2 | XP_006239500.1 | 2, 3 | 973 | |||
XM_008764198.1 | XP_008762420.1 | 2, 3 | 973 | |||
XM_008764199.1 | XP_008762421.1 | 2, 3 | 1390 | |||
XM_008764200.1 | XP_008762422.1 | 2, 3 | 973 | |||
XM_008764201.1 | XP_008762423.1 | 2, 3 | 974 | |||
GenBank mRNA | AW920332.1 | - | 2, 3 | 249 | ||
U44091.1 | - | 2, 3 | 906 |
