siRNA Details
Gene Symbol: Cdk5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053891.1 | NP_446343.1 | 1 | 333 | ||
GenBank mRNA | U50707.1 | - | 1 | 333 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdk5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053891.1 | NP_446343.1 | 1 | 333 | ||
GenBank mRNA | U50707.1 | - | 1 | 333 |