siRNA Details
Gene Symbol: Jdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053894.1 | NP_446346.1 | 3 | 685 | ||
XM_008764734.1 | XP_008762956.1 | 4 | 1255 | |||
XM_008764735.1 | XP_008762957.1 | 4 | 582 | |||
GenBank mRNA | U53449.1 | - | 3 | 685 |
