siRNA Details
Gene Symbol: Il16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105749.1 | NP_001099219.1 | 14 | 3163 | ||
XM_006229488.2 | XP_006229550.1 | 14 | 4810 | |||
XM_006229489.2 | XP_006229551.1 | 14 | 4809 | |||
XM_008759599.1 | XP_008757821.1 | 14 | 6626 | |||
XM_008759600.1 | XP_008757822.1 | 15 | 3182 | |||
XM_008759601.1 | XP_008757823.1 | 2 | 1107 |
