siRNA Details
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057205.2 | NP_476553.1 | 4 | 203 | ||
XM_006236936.2 | XP_006236998.1 | 3 | 186 | |||
GenBank mRNA | AF336829.1 | - | 4 | 212 | ||
BC081743.1 | - | 4 | 203 | 7124631 | ||
FQ234407.1 | - | 3 | 162 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011666, NM_001111106, AK166655, AK168859, BC002002, and others.
Gene Ontology Categories:
