siRNA Details
Gene Symbol: Bmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080407.1 | NP_536332.1 | 12 | 2640 | ||
XM_006244951.2 | XP_006245013.1 | 13 | 3126 | |||
XM_006244952.2 | XP_006245014.1 | 13 | 3126 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007561, AK148180, AK138715, AF003942, U78048.
Gene Ontology Categories:
