siRNA Details
Gene Symbol: Faim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080895.1 | NP_543171.1 | 249 | |||
XM_006243616.2 | XP_006243678.1 | 396 | ||||
XM_006243617.2 | XP_006243679.1 | 490 | ||||
XM_006243619.2 | XP_006243681.1 | 433 | ||||
XM_006243620.2 | XP_006243682.1 | 517 | ||||
XM_008766474.1 | XP_008764696.1 | 606 | ||||
GenBank mRNA | AF412334.1 | - | 249 | |||
AF467453.1 | - | 230 |
Gene Symbol: LOC100362113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729959.3 | XP_002730005.1 | 4 | 396 | ||
XM_006243611.2 | XP_006243673.1 | 4 | 490 | |||
XM_006243613.2 | XP_006243675.1 | 3 | 337 | |||
XM_006243614.2 | XP_006243676.1 | 3 | 433 | |||
XM_006243615.2 | XP_006243677.1 | 3 | 517 | |||
XM_008766582.1 | XP_008764804.1 | 4 | 606 |
