siRNA Details
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009953.2 | NP_001009953.2 | 2 | 331 | ||
NM_080909.2 | NP_543185.2 | 3 | 2505 | |||
XM_006230649.2 | XP_006230711.1 | 4 | 2577 | |||
XM_006230650.2 | XP_006230712.1 | 2 | 301 | |||
GenBank mRNA | AF442357.1 | - | 2 | 335 | ||
AF442358.1 | - | 2 | 335 | |||
AY164698.1 | - | 2 | 335 | |||
AY496443.1 | - | 3 | 2505 | |||
BC062068.1 | - | 2 | 329 | |||
BC107448.1 | - | 2 | 331 | |||
BC129077.1 | - | 2 | 339 |
