siRNA Details
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130409.2 | NP_569093.2 | 2 | 160 | ||
XM_008769531.1 | XP_008767753.1 | 2 | 357 | |||
XM_008769532.1 | XP_008767754.1 | 2 | 356 | |||
XM_008769533.1 | XP_008767755.1 | 2 | 357 | |||
GenBank mRNA | AJ320522.1 | - | 134 | |||
BC083174.1 | - | 169 | 7096787 | |||
CB816813.1 | - | 2 | 160 |
