siRNA Details
Gene Symbol: Hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191845.1 | NP_001178774.1 | 5 | 512 | ||
GenBank mRNA | AY059383.1 | - | 2 | 190 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191845.1 | NP_001178774.1 | 5 | 512 | ||
GenBank mRNA | AY059383.1 | - | 2 | 190 |