siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 2 | 709 | ||
NM_001270590.1 | NP_001257519.1 | 2 | 394 | |||
NM_001270591.1 | NP_001257520.1 | 3 | 172 | |||
NM_001270592.1 | NP_001257521.1 | 3 | 169 | |||
NM_001270593.1 | NP_001257522.1 | 2 | 106 | |||
NM_001270594.1 | NP_001257523.1 | 2 | 103 | |||
NM_134390.3 | NP_599217.1 | 2 | 712 | |||
XM_006236455.2 | XP_006236517.1 | 2 | 95 | |||
GenBank mRNA | AF370882.1 | - | 1 | 65 | ||
BC059116.1 | - | 2 | 211 | 6920949 | ||
FM070640.1 | - | 2 | 93 | |||
FM086181.1 | - | 3 | 156 | |||
FN804490.1 | - | 3 | 155 | |||
FQ209634.1 | - | 2 | 100 | |||
FQ212987.1 | - | 2 | 101 | |||
FQ219149.1 | - | 3 | 156 | |||
FQ219532.1 | - | 3 | 156 | |||
FQ219576.1 | - | 2 | 100 | |||
FQ220000.1 | - | 2 | 285 | |||
FQ233370.1 | - | 3 | 166 | |||
FQ235102.1 | - | 2 | 118 |
