siRNA Details
Gene Symbol: Micu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134396.2 | NP_599223.2 | 7 | 647 | ||
GenBank mRNA | BQ207114.1 | - | 2 | 91 | ||
DY317464.1 | - | 5 | 297 | |||
EV765275.1 | - | 7 | 654 |
